siRNA Details
| Gene Symbol: LMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006769.3 | NP_006760.1 | 5 | 1372 | ||
| XM_005271291.3 | XP_005271348.1 | 5 | 3542 | |||
| GenBank mRNA | AK292578.1 | - | 5 | 974 | ||
| BC003600.1 | - | 5 | 735 | 3355972 | ||
| BC017673.1 | - | 5 | 860 | 3893112 | ||
| BC065818.1 | - | 5 | 666 | 6196307 | ||
| U24576.1 | - | 5 | 1372 | |||

LMO4
Genomic Map