siRNA Details
| Gene Symbol: ZNF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006961.3 | NP_008892.2 | 6 | 561 | ||
| GenBank mRNA | AK055567.1 | - | 5 | 442 | ||
| AK057330.1 | - | 5 | 736 | |||
| AK292260.1 | - | 6 | 759 | |||
| BC047091.1 | - | 6 | 548 | 5270853 | ||

ZNF19
Genomic Map