siRNA Details
| Gene Symbol: RNF113A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006978.2 | NP_008909.1 | 1 | 937 | ||
| GenBank mRNA | AF086291.1 | - | 1 | 229 | ||
| AK314778.1 | - | 1 | 883 | |||
| BC000832.1 | - | 1 | 780 | 3455861 | ||
| BC020556.1 | - | 1 | 823 | 3447759 | ||
| BT007175.1 | - | 1 | 722 | |||
| X98253.1 | - | 1 | 932 | |||

RNF113A
Genomic Map