siRNA Details
| Gene Symbol: ZNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007147.3 | NP_009078.1 | 5 | 1051 | ||
| XM_017027249.1 | XP_016882738.1 | 5 | 1069 | |||
| GenBank mRNA | AK292687.1 | - | 5 | 984 | ||
| BC052997.1 | - | 5 | 983 | 6173867 | ||
| D50419.1 | - | 5 | 977 | |||

ZNF175
Genomic Map