siRNA Details
Gene Symbol: ZNF195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209664.1 | - | 1 | 10397 | ||
AF003540.1 | - | 2343 | ||||
AL833722.1 | - | 4 | 2177 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209664.1 | - | 1 | 10397 | ||
AF003540.1 | - | 2343 | ||||
AL833722.1 | - | 4 | 2177 |