siRNA Details
| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003855.3 | NP_003846.1 | 3 | 434 | ||
| XM_005264039.3 | XP_005264096.1 | 2 | 316 | |||
| XM_005264040.3 | XP_005264097.1 | 2 | 316 | |||
| XM_011512099.1 | XP_011510401.1 | 2 | 316 | |||
| XM_017005181.1 | XP_016860670.1 | 2 | 316 | |||
| GenBank mRNA | AY192162.1 | - | 2 | 288 | ||
| BC069575.1 | - | 2 | 312 | 7262302 | ||
| BC093975.1 | - | 2 | 312 | 7939820 | ||
| BC093977.1 | - | 2 | 312 | 7939822 | ||
| BC143403.1 | - | 2 | 312 | 9051910 | ||
| U43672.1 | - | 2 | 312 | |||

IL18R1
Genomic Map