siRNA Details
| Gene Symbol: ZNF225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321685.1 | NP_001308614.1 | 6 | 2428 | ||
| NM_013362.3 | NP_037494.2 | 5 | 2263 | |||
| XM_005259223.3 | XP_005259280.1 | 2 | 1809 | |||
| XM_011527285.2 | XP_011525587.1 | 5 | 2283 | |||
| XM_011527286.2 | XP_011525588.1 | 5 | 2927 | |||
| GenBank mRNA | AF187991.1 | - | 5 | 2237 | ||
| AK223477.1 | - | 5 | 2182 | |||
| AK292437.1 | - | 5 | 2267 | |||
| AK302106.1 | - | 4 | 1979 | |||
| AK313301.1 | - | 4 | 2270 | |||
| BC108912.2 | - | 4 | 2060 | 40027157 | ||

ZNF225
Genomic Map