siRNA Details
| Gene Symbol: ZNF112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083335.1 | NP_001076804.1 | 5 | 547 | ||
| NM_013380.3 | NP_037512.3 | 4 | 498 | |||
| GenBank mRNA | AK023099.1 | - | 4 | 468 | ||
| AK302966.1 | - | 5 | 605 | |||
| AU130692.1 | - | 4 | 468 | |||
| BC101753.1 | - | 4 | 442 | 8069259 | ||
| BC117223.1 | - | 4 | 442 | 40125774 | ||

ZNF112
Genomic Map