siRNA Details
| Gene Symbol: ZNF112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083335.1 | NP_001076804.1 | 5 | 3018 | ||
| NM_013380.3 | NP_037512.3 | 4 | 2969 | |||
| GenBank mRNA | AF198358.1 | - | 5 | 2961 | ||
| AK023099.1 | - | 4 | 2939 | |||
| AK026230.1 | - | 1 | 658 | |||
| AK130371.1 | - | 1 | 1471 | |||
| AK302966.1 | - | 5 | 3076 | |||
| AL122078.1 | - | 1 | 485 | |||

ZNF112
Genomic Map