siRNA Details
| Gene Symbol: ZNF224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321645.1 | NP_001308574.1 | 6 | 2234 | ||
| NM_013398.3 | NP_037530.2 | 6 | 2192 | |||
| XM_017027261.1 | XP_016882750.1 | 5 | 2268 | |||
| GenBank mRNA | AF067164.1 | - | 1 | 2709 | ||
| AF187990.2 | - | 6 | 2177 | |||
| AK292500.1 | - | 6 | 2172 | |||
| AY148489.1 | - | 1 | 1620 | |||
| BC051902.1 | - | 6 | 2129 | 6155240 | ||

ZNF224
Genomic Map