siRNA Details
| Gene Symbol: ZNF26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 1984 | ||
| NM_001256280.1 | NP_001243209.1 | 3 | 1789 | |||
| NM_019591.3 | NP_062537.2 | 4 | 1885 | |||
| XM_005266182.3 | XP_005266239.1 | 5 | 1972 | |||
| XM_005266183.3 | XP_005266240.1 | 6 | 2175 | |||
| XM_011534829.1 | XP_011533131.1 | 5 | 2088 | |||
| XM_017019920.1 | XP_016875409.1 | 6 | 2608 | |||
| XM_017019921.1 | XP_016875410.1 | 6 | 2071 | |||
| XM_017019922.1 | XP_016875411.1 | 5 | 2957 | |||
| XM_017019923.1 | XP_016875412.1 | 5 | 2888 | |||
| GenBank mRNA | AK000762.1 | - | 1 | 1088 | ||
| AK122941.1 | - | 3 | 3998 | |||
| AK225296.1 | - | 1 | 1088 | |||
| AK291243.1 | - | 4 | 1852 | |||
| AL833057.1 | - | 1 | 662 | |||
| BC008211.1 | - | 1 | 625 | |||
| BC046206.2 | - | 4 | 1786 | 6176427 | ||
| BT006963.1 | - | 1 | 390 | |||
| EU668356.1 | - | 4 | 1763 | |||
| X52351.1 | - | 1 | 33 | |||

ZNF26
Genomic Map