siRNA Details
| Gene Symbol: TUFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126337.1 | NP_001119809.1 | 10 | 976 | ||
| NM_001301317.1 | NP_001288246.1 | 12 | 1186 | |||
| NM_020127.2 | NP_064512.1 | 11 | 1051 | |||
| XM_011509961.2 | XP_011508263.1 | 9 | 885 | |||
| XM_017002223.1 | XP_016857712.1 | 11 | 1127 | |||
| XM_017002224.1 | XP_016857713.1 | 10 | 1248 | |||
| GenBank mRNA | AF254260.1 | - | 11 | 1051 | ||
| AK075001.1 | - | 10 | 973 | |||
| AK223161.1 | - | 11 | 1047 | |||
| AK298066.1 | - | 12 | 1186 | |||
| AK299925.1 | - | 10 | 964 | |||
| AK315414.1 | - | 11 | 1047 | |||
| AL136917.1 | - | 11 | 1039 | |||
| BC002933.1 | - | 10 | 944 | 3956873 | ||
| BC008301.1 | - | 11 | 1031 | 3506004 | ||

TUFT1
Genomic Map