siRNA Details
| Gene Symbol: HIST1H2AJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021066.2 | NP_066544.1 | 1 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AJ| Gene Symbol: HIST1H2AJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021066.2 | NP_066544.1 | 1 | 399 | ||