siRNA Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021964.2 | NP_068799.2 | 5 | 902 | ||
| GenBank mRNA | AF039019.1 | - | 4 | 627 | ||
| AF432210.1 | - | 5 | 718 | |||
| AJ236885.1 | - | 5 | 807 | |||
| AK314203.1 | - | 5 | 837 | |||
| AL833616.1 | - | 5 | 739 | |||
| BC035591.1 | - | 5 | 813 | |||
| BC050260.1 | - | 6 | 978 | 4423572 | ||
| BQ441085.1 | - | 2 | 156 | |||
| DQ090088.1 | - | 2 | 412 | |||
| L04282.1 | - | 5 | 806 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011749, AK036835, CK634070, U80078, BC026144, and others.

ZNF148
Genomic Map