siRNA Details
| Gene Symbol: AKAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003886.2 | NP_003877.2 | 5 | 1720 | ||
| NM_139289.1 | NP_647450.1 | 5 | 1693 | |||
| GenBank mRNA | AF072756.1 | - | 5 | 1748 | ||
| BC033007.1 | - | 5 | 1713 | |||
| BC126250.1 | - | 5 | 1719 | 8991966 | ||
| BC126252.1 | - | 5 | 1719 | 8991968 | ||

AKAP4
Genomic Map