siRNA Details
| Gene Symbol: EMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320049.1 | NP_001306978.1 | 5 | 687 | ||
| NM_006331.7 | NP_006322.4 | 6 | 837 | |||
| GenBank mRNA | AK291597.1 | - | 6 | 702 | ||
| AK307177.1 | - | 3 | 470 | |||
| BC055314.1 | - | 6 | 729 | 4718702 | ||
| BP315329.1 | - | 5 | 555 | |||
| U72514.1 | - | 6 | 680 | |||

EMG1
Genomic Map