siRNA Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300950.1 | NP_001287879.1 | 2 | 341 | ||
| NM_006530.3 | NP_006521.1 | 2 | 341 | |||
| GenBank mRNA | AJ245746.1 | - | 2 | 289 | ||
| AK001413.1 | - | 2 | 114 | |||
| BC000994.2 | - | 2 | 202 | 2899965 | ||
| CA775118.1 | - | 2 | 241 | |||
| DN915967.1 | - | 2 | 342 | |||
| U61384.1 | - | 2 | 299 | |||

YEATS4
Genomic Map