siRNA Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300950.1 | NP_001287879.1 | 4 | 602 | ||
| NM_006530.3 | NP_006521.1 | 6 | 764 | |||
| GenBank mRNA | AJ245746.1 | - | 6 | 712 | ||
| AK001413.1 | - | 6 | 537 | |||
| BC000994.2 | - | 6 | 625 | 2899965 | ||
| CA775118.1 | - | 4 | 502 | |||
| U61384.1 | - | 6 | 722 | |||

YEATS4
Genomic Map