siRNA Details
| Gene Symbol: SNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317192.1 | NP_001304121.1 | 5, 6 | 527 | ||
| NM_001317193.1 | NP_001304122.1 | 4, 5 | 476 | |||
| NM_001317194.1 | NP_001304123.1 | 7, 8 | 617 | |||
| NM_007241.3 | NP_009172.2 | 5, 6 | 527 | |||
| XM_017024075.1 | XP_016879564.1 | 6, 7 | 548 | |||
| GenBank mRNA | AF156102.1 | - | 5, 6 | 476 | ||
| BC008976.1 | - | 5, 6 | 477 | 3452153 | ||
| BC038830.1 | - | 5, 6 | 501 | 3900806 | ||
| BE394098.1 | - | 7, 8 | 519 | |||
| BE818533.1 | - | 4, 5 | 330 | |||
| BP342490.1 | - | 5, 6 | 527 | |||

SNF8
Genomic Map