siRNA Details
| Gene Symbol: DEXI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014015.3 | NP_054734.2 | 2 | 858 | ||
| XM_011522455.2 | XP_011520757.1 | 3 | 240 | |||
| XM_017023171.1 | XP_016878660.1 | 3 | 1626 | |||
| GenBank mRNA | AF108145.2 | - | 2 | 474 | ||
| AF129929.1 | - | 2 | 370 | |||
| BC001083.2 | - | 2 | 594 | 2958243 | ||
| BC037569.1 | - | 2 | 858 | 5227606 | ||
| BC052989.1 | - | 2 | 591 | 6194689 | ||
| BC071683.1 | - | 2 | 757 | 5190179 | ||

DEXI
Genomic Map