siRNA Details
| Gene Symbol: C16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014117.2 | NP_054836.2 | 4 | 1830 | ||
| GenBank mRNA | AK123266.1 | - | 2 | 1094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf72| Gene Symbol: C16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014117.2 | NP_054836.2 | 4 | 1830 | ||
| GenBank mRNA | AK123266.1 | - | 2 | 1094 | ||