siRNA Details
| Gene Symbol: UTP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014503.2 | NP_055318.2 | 14 | 1769 | ||
| GenBank mRNA | AJ006778.1 | - | 14 | 1757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UTP20| Gene Symbol: UTP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014503.2 | NP_055318.2 | 14 | 1769 | ||
| GenBank mRNA | AJ006778.1 | - | 14 | 1757 | ||