siRNA Details
| Gene Symbol: KIAA0101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029989.2 | NP_001025160.1 | 3 | 793 | ||
| NM_014736.5 | NP_055551.1 | 4 | 956 | |||
| NR_109934.1 | 4 | 1012 | ||||
| GenBank mRNA | BC005832.2 | - | 4 | 876 | 2988995 | |

KIAA0101
Genomic Map