siRNA Details
| Gene Symbol: ARSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267727.1 | NP_001254656.1 | 12 | 2203 | ||
| NM_014960.4 | NP_055775.2 | 12 | 2374 | |||
| XM_005257170.3 | XP_005257227.1 | 12 | 2387 | |||
| XM_006721777.3 | XP_006721840.2 | 13 | 2482 | |||
| XM_011524535.2 | XP_011522837.1 | 13 | 2480 | |||
| XM_011524536.2 | XP_011522838.1 | 13 | 2310 | |||
| XM_011524537.1 | XP_011522839.1 | 13 | 2231 | |||
| XM_011524538.2 | XP_011522840.1 | 13 | 2482 | |||
| XM_011524540.2 | XP_011522842.1 | 13 | 2502 | |||
| XM_011524545.2 | XP_011522847.1 | 11 | 2304 | |||
| XM_011524546.2 | XP_011522848.1 | 8 | 1153 | |||
| XM_017024360.1 | XP_016879849.1 | 13 | 2320 | |||
| XM_017024361.1 | XP_016879850.1 | 12 | 2387 | |||
| XM_017024362.1 | XP_016879851.1 | 12 | 2326 | |||
| XM_017024363.1 | XP_016879852.1 | 12 | 2232 | |||
| XM_017024364.1 | XP_016879853.1 | 12 | 2228 | |||
| XM_017024365.1 | XP_016879854.1 | 12 | 2216 | |||
| XM_017024368.1 | XP_016879857.1 | 11 | 2108 | |||
| GenBank mRNA | AB023218.1 | - | 11 | 2036 | ||
| AK311327.1 | - | 3 | 777 | |||
| AY358380.1 | - | 12 | 2374 | |||
| BC012375.1 | - | 11 | 1617 | 3882163 | ||

ARSG
Genomic Map