siRNA Details
| Gene Symbol: TBC1D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015079.5 | NP_055894.6 | 3 | 636 | ||
| NM_144572.1 | NP_653173.1 | 3 | 636 | |||
| XM_011521387.2 | XP_011519689.1 | 3 | 692 | |||
| XM_017022022.1 | XP_016877511.1 | 3 | 512 | |||
| GenBank mRNA | AB028978.1 | - | 3 | 499 | ||
| BC140766.1 | - | 2 | 281 | 9021634 | ||
| BC144482.1 | - | 2 | 281 | 9053009 | ||
| BC152465.1 | - | 3 | 499 | 8862601 | ||

TBC1D2B
Genomic Map