siRNA Details
| Gene Symbol: ZCCHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015144.2 | NP_055959.1 | 12 | 1651 | ||
| XM_005255858.3 | XP_005255915.2 | 12 | 1842 | |||
| XM_017023082.1 | XP_016878571.1 | 12 | 2601 | |||
| GenBank mRNA | AB011151.2 | - | 12 | 1509 | ||
| AB030243.1 | - | 12 | 1659 | |||
| AK289682.1 | - | 12 | 1842 | |||
| AL117532.1 | - | 9 | 1274 | |||
| BC101478.1 | - | 12 | 1658 | 8068984 | ||

ZCCHC14
Genomic Map