siRNA Details
| Gene Symbol: DMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174116.1 | NP_001167587.1 | 7 | 942 | ||
| NM_001174117.1 | NP_001167588.1 | 7 | 942 | |||
| NM_015263.3 | NP_056078.2 | 7 | 942 | |||
| XM_005254255.1 | XP_005254312.1 | 7 | 1005 | |||
| XM_005254256.1 | XP_005254313.1 | 7 | 1005 | |||
| XM_011521402.2 | XP_011519704.1 | 7 | 1005 | |||
| XM_017022034.1 | XP_016877523.1 | 7 | 678 | |||
| GenBank mRNA | AF389880.1 | - | 7 | 924 | ||
| BC140781.1 | - | 7 | 807 | 9021649 | ||
| BC144539.1 | - | 7 | 807 | 9053066 | ||

DMXL2
Genomic Map