siRNA Details
| Gene Symbol: DMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174116.1 | NP_001167587.1 | 8 | 1076 | ||
| NM_001174117.1 | NP_001167588.1 | 8 | 1076 | |||
| NM_015263.3 | NP_056078.2 | 8 | 1076 | |||
| XM_005254255.1 | XP_005254312.1 | 8 | 1139 | |||
| XM_005254256.1 | XP_005254313.1 | 8 | 1139 | |||
| XM_011521402.2 | XP_011519704.1 | 8 | 1139 | |||
| XM_017022034.1 | XP_016877523.1 | 8 | 812 | |||
| GenBank mRNA | AF389880.1 | - | 8 | 1058 | ||
| BC140781.1 | - | 8 | 941 | 9021649 | ||
| BC144539.1 | - | 8 | 941 | 9053066 | ||

DMXL2
Genomic Map