siRNA Details
| Gene Symbol: METTL21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015433.2 | NP_056248.2 | 3 | 1024 | ||
| NM_206914.1 | NP_996797.1 | 4 | 1163 | |||
| GenBank mRNA | AF455816.1 | - | 3 | 1024 | ||
| AK024983.1 | - | 4 | 1051 | |||
| AK292276.1 | - | 3 | 968 | |||
| AL050100.1 | - | 4 | 1085 | |||
| BC016395.1 | - | 3 | 1024 | 3882414 | ||
| BC099841.1 | - | 3 | 999 | 40010149 | ||
| BC103501.1 | - | 3 | 999 | 40010152 | ||

METTL21B
Genomic Map