siRNA Details
| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286487.1 | NP_001273416.1 | 7 | 772 | ||
| NM_015497.4 | NP_056312.2 | 8 | 833 | |||
| XM_005254287.1 | XP_005254344.1 | 8 | 684 | |||
| XM_006720468.2 | XP_006720531.1 | 8 | 689 | |||
| XM_011521443.2 | XP_011519745.1 | 7 | 601 | |||
| XM_011521444.2 | XP_011519746.1 | 8 | 684 | |||
| XM_017022067.1 | XP_016877556.1 | 8 | 689 | |||
| GenBank mRNA | AF132733.1 | - | 8 | 689 | ||
| AK074870.1 | - | 8 | 682 | |||
| AK075403.1 | - | 8 | 719 | |||
| AK289591.1 | - | 8 | 682 | |||
| AK290150.1 | - | 8 | 668 | |||
| AK290484.1 | - | 7 | 616 | |||
| AK297816.1 | - | 8 | 678 | |||
| AK309915.1 | - | 8 | 682 | |||
| AK309954.1 | - | 7 | 622 | |||

TMEM87A
Genomic Map