siRNA Details
| Gene Symbol: SLC25A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282195.1 | NP_001269124.1 | 4 | 564 | ||
| NM_001282196.1 | NP_001269125.1 | 4 | 555 | |||
| NM_001282197.1 | NP_001269126.1 | 4 | 555 | |||
| NM_001282198.1 | NP_001269127.1 | 3 | 317 | |||
| NR_104107.1 | 3 | 317 | ||||
| XM_011531402.2 | XP_011529704.1 | 4 | 566 | |||
| XM_011531403.1 | XP_011529705.1 | 3 | 280 | |||
| XM_017029936.1 | XP_016885425.1 | 3 | 280 | |||
| XM_017029937.1 | XP_016885426.1 | 4 | 566 | |||
| XM_017029938.1 | XP_016885427.1 | 4 | 561 | |||
| XM_017029939.1 | XP_016885428.1 | 4 | 530 | |||
| XM_017029941.1 | XP_016885430.1 | 4 | 539 | |||
| GenBank mRNA | AF078544.1 | - | 3 | 560 | ||
| AF155809.1 | - | 3 | 303 | |||
| AF155810.1 | - | 3 | 294 | |||
| AF155811.1 | - | 3 | 294 | |||
| AK296155.1 | - | 3 | 247 | |||
| AK297499.1 | - | 4 | 498 | |||
| AK304649.1 | - | 3 | 297 | |||
| AK309433.1 | - | 4 | 489 | |||
| AL520510.3 | - | 3 | 267 | |||
| AY358099.1 | - | 3 | 312 | |||
| BC119666.2 | - | 3 | 419 | 40116117 | ||
| BC119667.1 | - | 3 | 419 | 40116118 | ||
| DB485834.2 | - | 3 | 317 | |||

SLC25A14
Genomic Map