siRNA Details
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042399.1 | NP_001035858.1 | 7 | 1358 | ||
| NM_016122.2 | NP_057206.2 | 8 | 1411 | |||
| XM_006719437.2 | XP_006719500.1 | 7 | 1324 | |||
| XM_011538424.1 | XP_011536726.1 | 9 | 1457 | |||
| XM_011538427.2 | XP_011536729.1 | 8 | 1183 | |||
| XM_011538429.2 | XP_011536731.1 | 4 | 654 | |||
| XM_017019385.1 | XP_016874874.1 | 8 | 1405 | |||
| XM_017019386.1 | XP_016874875.1 | 8 | 1405 | |||
| XM_017019387.1 | XP_016874876.1 | 7 | 1162 | |||
| XM_017019388.1 | XP_016874877.1 | 7 | 3027 | |||
| XM_017019389.1 | XP_016874878.1 | 9 | 1237 | |||
| GenBank mRNA | AF155115.1 | - | 8 | 1127 | ||
| AK056316.1 | - | 7 | 1254 | |||
| BC107746.1 | - | 5 | 672 | |||
| BC125086.1 | - | 7 | 1048 | |||
| BC125087.1 | - | 8 | 961 | 40117807 | ||

CEP83
Genomic Map