siRNA Details
| Gene Symbol: GPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164372.1 | NP_001157844.1 | 8 | 1346 | ||
| NM_001164373.1 | NP_001157845.1 | 8 | 1255 | |||
| NM_016301.3 | NP_057385.3 | 8 | 1225 | |||
| XM_005253896.4 | XP_005253953.3 | 9 | 1680 | |||
| XM_017019394.1 | XP_016874883.1 | 8 | 1160 | |||
| GenBank mRNA | AF117229.1 | - | 8 | 1175 | ||
| AK223209.1 | - | 8 | 1197 | |||
| BC008416.1 | - | 8 | 1200 | 4048678 | ||
| BC031024.1 | - | 8 | 1201 | 4695834 | ||
| BX537973.1 | - | 8 | 1236 | |||

GPN3
Genomic Map