siRNA Details
| Gene Symbol: GLTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016433.3 | NP_057517.1 | 3 | 294 | ||
| XM_011538434.2 | XP_011536736.1 | 4 | 586 | |||
| GenBank mRNA | AF209704.1 | - | 3 | 190 | ||
| AK313457.1 | - | 3 | 183 | |||
| AY372530.1 | - | 3 | 180 | |||
| AY372531.1 | - | 3 | 180 | |||
| AY372532.1 | - | 3 | 209 | |||
| BC009932.2 | - | 3 | 294 | 3959556 | ||
| JN039379.1 | - | 4 | 519 | |||

GLTP
Genomic Map