siRNA Details
| Gene Symbol: EMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286420.1 | NP_001273349.1 | 3 | 451 | ||
| NM_016454.3 | NP_057538.1 | 3 | 451 | |||
| GenBank mRNA | AF151018.1 | - | 3 | 404 | ||
| AK075227.1 | - | 3 | 430 | |||
| AK290524.1 | - | 3 | 435 | |||
| AY336092.1 | - | 3 | 391 | |||
| BC002583.2 | - | 3 | 416 | 3161718 | ||
| KU178640.1 | - | 3 | 333 | |||
| KU178641.1 | - | 3 | 333 | |||

EMC4
Genomic Map