siRNA Details
| Gene Symbol: MYOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003970.3 | NP_003961.3 | 27 | 3508 | ||
| XM_006716237.1 | XP_006716300.1 | 27 | 3473 | |||
| GenBank mRNA | AK297890.1 | - | 14 | 1826 | ||
| BC052969.1 | - | 27 | 3461 | 6205359 | ||
| BP260135.1 | - | 2 | 181 | |||
| BP260324.1 | - | 2 | 185 | |||
| DA900083.1 | - | 6 | 498 | |||
| X69089.1 | - | 27 | 3418 | |||

MYOM2
Genomic Map