siRNA Details
| Gene Symbol: TTC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017775.3 | NP_060245.3 | 2, 3 | 784 | ||
| XM_017024801.1 | XP_016880290.1 | 2, 3 | 697 | |||
| XM_017024802.1 | XP_016880291.1 | 2, 3 | 697 | |||
| GenBank mRNA | AK056878.1 | - | 2, 3 | 696 | ||
| AK297783.1 | - | 2, 3 | 340 | |||
| BC011698.2 | - | 2, 3 | 323 | |||
| BC073796.1 | - | 2, 3 | 327 | |||
| BC105128.1 | - | 2, 3 | 784 | 8144131 | ||
| BC112107.1 | - | 2, 3 | 784 | 8327575 | ||

TTC19
Genomic Map