siRNA Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316943.1 | NP_001303872.1 | 3 | 852 | ||
| NM_017851.5 | NP_060321.3 | 3 | 852 | |||
| XM_006720589.2 | XP_006720652.1 | 3 | 893 | |||
| XM_006720590.2 | XP_006720653.1 | 3 | 893 | |||
| XM_006720592.2 | XP_006720655.1 | 3 | 836 | |||
| XM_011521742.2 | XP_011520044.1 | 2 | 757 | |||
| XM_017022386.1 | XP_016877875.1 | 3 | 836 | |||
| XM_017022387.1 | XP_016877876.1 | 2 | 757 | |||
| GenBank mRNA | AK058010.1 | - | 3 | 436 | ||
| BC006389.1 | - | 3 | 698 | 4127509 | ||
| BC031074.1 | - | 3 | 501 | 5274010 | ||

PARP16
Genomic Map