siRNA Details
| Gene Symbol: TESC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168325.1 | NP_001161797.1 | 2 | 250 | ||
| NM_017899.3 | NP_060369.3 | 2 | 250 | |||
| NR_031766.2 | 2 | 250 | ||||
| XM_011538503.1 | XP_011536805.1 | 2 | 65 | |||
| XM_017019532.1 | XP_016875021.1 | 2 | 294 | |||
| XM_017019533.1 | XP_016875022.1 | 2 | 294 | |||
| GenBank mRNA | AF443207.1 | - | 2 | 207 | ||
| AK000614.1 | - | 2 | 293 | |||
| BC015221.1 | - | 2 | 207 | 3885280 | ||
| BQ434057.1 | - | 2 | 183 | |||
| BQ949772.1 | - | 2 | 204 | |||
| DB452909.1 | - | 2 | 250 | |||

TESC
Genomic Map