siRNA Details
| Gene Symbol: SCYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317784.1 | NP_001304713.1 | 5 | 783 | ||
| NM_017988.5 | NP_060458.3 | 5 | 935 | |||
| XM_017019618.1 | XP_016875107.1 | 5 | 783 | |||
| XM_017019619.1 | XP_016875108.1 | 5 | 935 | |||
| XM_017019620.1 | XP_016875109.1 | 6 | 922 | |||
| GenBank mRNA | AB037781.1 | - | 2 | 89 | ||
| AK000936.1 | - | 4 | 541 | |||
| AK125091.1 | - | 5 | 920 | |||
| AK292980.1 | - | 5 | 709 | |||
| BC063798.1 | - | 5 | 548 | 6473586 | ||

SCYL2
Genomic Map