siRNA Details
| Gene Symbol: SLC38A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308384.1 | NP_001295313.1 | 8 | 2531 | ||
| NM_018231.2 | NP_060701.1 | 12 | 2466 | |||
| XM_017023393.1 | XP_016878882.1 | 11 | 2947 | |||
| XM_017023394.1 | XP_016878883.1 | 9 | 2039 | |||
| XM_017023395.1 | XP_016878884.1 | 11 | 2242 | |||
| XM_017023396.1 | XP_016878885.1 | 10 | 2094 | |||
| GenBank mRNA | AK001677.1 | - | 12 | 2459 | ||
| BC001961.1 | - | 11 | 2415 | 3510557 | ||

SLC38A7
Genomic Map