siRNA Details
| Gene Symbol: SLC47A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018242.2 | NP_060712.2 | 12, 13 | 1198 | ||
| GenBank mRNA | AK001709.1 | - | 12, 13 | 1198 | ||
| AK222625.1 | - | 12, 13 | 1202 | |||
| AK293196.1 | - | 5, 6 | 491 | |||
| AK298254.1 | - | 3, 4 | 440 | |||
| AK301854.1 | - | 11, 12 | 1155 | |||
| AK304647.1 | - | 4, 5 | 377 | |||
| AK310270.1 | - | 10, 11 | 877 | |||
| BC010661.2 | - | 12, 13 | 1187 | 3874616 | ||
| BC050592.1 | - | 12, 13 | 1172 | |||

SLC47A1
Genomic Map