siRNA Details
| Gene Symbol: TMEM100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099640.1 | NP_001093110.1 | 4 | 1629 | ||
| NM_018286.2 | NP_060756.2 | 2 | 1131 | |||
| XM_017024814.1 | XP_016880303.1 | 5 | 2986 | |||
| XM_017024815.1 | XP_016880304.1 | 3 | 1095 | |||
| XM_017024816.1 | XP_016880305.1 | 2 | 1147 | |||
| GenBank mRNA | AK001832.1 | - | 2 | 1048 | ||
| AK095175.1 | - | 4 | 1592 | |||
| BC010128.2 | - | 2 | 981 | 3952255 | ||

TMEM100
Genomic Map