siRNA Details
| Gene Symbol: ATIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004044.6 | NP_004035.2 | 5 | 535 | ||
| XM_017004187.1 | XP_016859676.1 | 5 | 458 | |||
| GenBank mRNA | AB062403.1 | - | 5 | 383 | ||
| AK313066.1 | - | 5 | 429 | |||
| BC008879.2 | - | 5 | 378 | 4300570 | ||
| D82348.1 | - | 5 | 423 | |||
| D89976.1 | - | 5 | 405 | |||
| DA043224.1 | - | 4 | 469 | |||
| EU794654.1 | - | 5 | 522 | |||
| U37436.1 | - | 4 | 345 | |||

ATIC
Genomic Map