siRNA Details
| Gene Symbol: LRRC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018509.3 | NP_060979.2 | 7 | 1764 | ||
| GenBank mRNA | AF119857.1 | - | 4 | 1122 | ||
| AK025328.1 | - | 7 | 1764 | |||
| AK130572.1 | - | 1 | 727 | |||
| BC052279.1 | - | 7 | 1682 | 4127952 | ||

LRRC59
Genomic Map