siRNA Details
| Gene Symbol: FAM64A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195228.1 | NP_001182157.1 | 5 | 1319 | ||
| NM_019013.2 | NP_061886.2 | 6 | 1364 | |||
| XM_017024778.1 | XP_016880267.1 | 4 | 1693 | |||
| GenBank mRNA | AK001018.1 | - | 5 | 1304 | ||
| AK001353.1 | - | 5 | 1279 | |||
| AK225613.1 | - | 6 | 1326 | |||
| AK225702.1 | - | 6 | 1324 | |||
| BC005004.1 | - | 5 | 1286 | 3533875 | ||
| BC013966.2 | - | 6 | 1310 | 2988318 | ||

FAM64A
Genomic Map