siRNA Details
| Gene Symbol: GPR162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014449.1 | NP_055264.1 | 5 | 1342 | ||
| NM_019858.1 | NP_062832.1 | 5 | 2344 | |||
| GenBank mRNA | AB209836.1 | - | 4 | 1751 | ||
| BC002353.2 | - | 5 | 1436 | 2821031 | ||
| BC007918.2 | - | 4 | 975 | |||
| BC080585.1 | - | 5 | 1323 | 3510624 | ||
| U47925.1 | - | 5 | 1342 | |||
| U47928.1 | - | 5 | 2344 | |||

GPR162
Genomic Map