siRNA Details
| Gene Symbol: LYZL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199951.2 | NP_001186880.1 | 3 | 516 | ||
| NM_020426.3 | NP_065159.1 | 3 | 630 | |||
| GenBank mRNA | AY359018.1 | - | 2 | 434 | ||
| AY742214.1 | - | 2 | 406 | |||
| BC054481.1 | - | 3 | 476 | 6620264 | ||
| BG195727.1 | - | 2 | 484 | |||
| GQ472214.1 | - | 3 | 476 | |||

LYZL6
Genomic Map