siRNA Details
| Gene Symbol: TLDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020947.3 | NP_065998.3 | 8 | 3117 | ||
| XM_005256074.3 | XP_005256131.1 | 8 | 3187 | |||
| XM_005256075.2 | XP_005256132.1 | 9 | 3315 | |||
| XM_006721240.2 | XP_006721303.1 | 9 | 3270 | |||
| XM_011523248.2 | XP_011521550.1 | 10 | 3414 | |||
| XM_017023510.1 | XP_016878999.1 | 10 | 3434 | |||
| XM_017023511.1 | XP_016879000.1 | 8 | 3189 | |||
| XM_017023512.1 | XP_016879001.1 | 9 | 3357 | |||
| GenBank mRNA | AB046829.2 | - | 3290 | |||
| AL137316.1 | - | 3 | 2062 | |||
| BC060844.1 | - | 9 | 3046 | 30342219 | ||

TLDC1
Genomic Map