siRNA Details
| Gene Symbol: IGDCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020962.2 | NP_066013.1 | 10 | 1973 | ||
| XM_011521845.2 | XP_011520147.1 | 10 | 2153 | |||
| XM_011521846.2 | XP_011520148.1 | 10 | 1777 | |||
| XM_011521847.1 | XP_011520149.1 | 10 | 1837 | |||
| XM_017022448.1 | XP_016877937.1 | 10 | 2155 | |||
| XM_017022449.1 | XP_016877938.1 | 10 | 2161 | |||
| GenBank mRNA | AB046848.1 | - | 4 | 1033 | ||
| AB052622.1 | - | 10 | 1973 | |||

IGDCC4
Genomic Map